Assay Details
Gene Symbol: LOC4343109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015790716.2 | XP_015646202.1 | 2 - 3 | 384 | 74 | |
GenBank mRNA | AK072251.1 | - | 2 - 3 | 323 | 74 | |
AK099317.1 | - | 2 - 3 | 337 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
