Assay Details
| Gene Symbol: LOC4343132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791831.2 | XP_015647317.1 | 7 - 8 | 1520 | 100 | |
| GenBank mRNA | AK287638.1 | - | 7 - 8 | 1490 | 100 | |
More Information
Set Membership: |
|

LOC4343132
Genomic Map