Assay Details
| Gene Symbol: LOC4343241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791778.1 | XP_015647264.1 | 5 - 6 | 818 | 128 | |
| GenBank mRNA | AK072721.1 | - | 5 - 6 | 818 | 128 | |
| HQ324821.1 | - | 5 - 6 | 808 | 128 | ||
More Information
Set Membership: |
|

LOC4343241
Genomic Map