Assay Details
| Gene Symbol: LOC4343241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791778.1 | XP_015647264.1 | 7 - 8 | 1084 | 118 | |
| GenBank mRNA | AK060926.1 | - | 2 - 3 | 243 | 118 | |
| HQ324821.1 | - | 7 - 8 | 1074 | 118 | ||
More Information
Set Membership: |
|

LOC4343241
Genomic Map