Assay Details
Gene Symbol: LOC4343445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791133.1 | XP_015646619.1 | 1 - 2 | 216 | 81 | |
GenBank mRNA | AF094774.1 | - | 1 - 2 | 116 | 81 | |
AK120697.1 | - | 1 - 2 | 136 | 81 | ||
BT129356.1 | - | 1 - 2 | 76 | 81 | ||
BT129377.1 | - | 1 - 2 | 130 | 81 | ||
BT130628.1 | - | 1 - 2 | 131 | 81 | ||
BT131551.1 | - | 1 - 2 | 134 | 81 | ||
BT131915.1 | - | 1 - 2 | 134 | 81 | ||
BT132390.1 | - | 1 - 2 | 129 | 81 | ||
BT132581.1 | - | 1 - 2 | 136 | 81 | ||
JN944373.1 | - | 1 - 2 | 120 | 81 | ||
KM982770.1 | - | 1 - 2 | 107 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015791133.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
