Assay Details
Gene Symbol: LOC4343445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791133.1 | XP_015646619.1 | 3 - 4 | 511 | 104 | |
GenBank mRNA | AF094774.1 | - | 3 - 4 | 411 | 104 | |
AK120697.1 | - | 3 - 4 | 431 | 104 | ||
BT129356.1 | - | 3 - 4 | 371 | 104 | ||
BT129377.1 | - | 3 - 4 | 425 | 104 | ||
BT130628.1 | - | 3 - 4 | 426 | 104 | ||
BT131551.1 | - | 3 - 4 | 429 | 104 | ||
BT131915.1 | - | 3 - 4 | 429 | 104 | ||
BT132390.1 | - | 3 - 4 | 424 | 104 | ||
BT132581.1 | - | 3 - 4 | 431 | 104 | ||
JN944373.1 | - | 3 - 4 | 415 | 104 | ||
KM982770.1 | - | 3 - 4 | 402 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
