Assay Details
| Gene Symbol: LOC4343445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791133.1 | XP_015646619.1 | 4 - 5 | 557 | 100 | |
| GenBank mRNA | AF094774.1 | - | 4 - 5 | 457 | 100 | |
| AK120697.1 | - | 4 - 5 | 477 | 100 | ||
| BT129356.1 | - | 4 - 5 | 417 | 100 | ||
| BT129377.1 | - | 4 - 5 | 471 | 100 | ||
| BT130628.1 | - | 4 - 5 | 472 | 100 | ||
| BT131551.1 | - | 4 - 5 | 475 | 100 | ||
| BT131915.1 | - | 4 - 5 | 475 | 100 | ||
| BT132390.1 | - | 4 - 5 | 470 | 100 | ||
| BT132581.1 | - | 4 - 5 | 477 | 100 | ||
| JN944373.1 | - | 4 - 5 | 461 | 100 | ||
| KM982770.1 | - | 4 - 5 | 448 | 100 | ||
More Information
Set Membership: |
|

LOC4343445
Genomic Map