Assay Details
Gene Symbol: LOC4343748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015790074.2 | XP_015645560.1 | 2 - 3 | 601 | 74 | |
GenBank mRNA | AK071404.1 | - | 2 - 3 | 568 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
