Assay Details
| Gene Symbol: LOC4343851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791998.2 | XP_015647484.1 | 5 - 6 | 578 | 144 | |
| GenBank mRNA | AJ011675.1 | - | 5 - 6 | 545 | 144 | |
| AK064704.1 | - | 5 - 6 | 558 | 144 | ||
More Information
Set Membership: |
|

LOC4343851
Genomic Map