Assay Details
Gene Symbol: LOC4344169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789524.2 | XP_015645010.1 | 3 - 4 | 749 | 92 | |
GenBank mRNA | DQ103591.1 | - | 3 - 4 | 373 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
