Assay Details
Gene Symbol: LOC4344169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789524.2 | XP_015645010.1 | 4 - 5 | 922 | 95 | |
GenBank mRNA | DQ103591.1 | - | 4 - 5 | 546 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
