Assay Details
Gene Symbol: LOC4344192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791726.1 | XP_015647212.1 | 13 - 14 | 2026 | 99 | |
GenBank mRNA | AK067082.1 | - | 13 - 14 | 1889 | 99 | |
KT199367.1 | - | 13 - 14 | 1790 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
