Assay Details
Gene Symbol: LOC4344192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791726.1 | XP_015647212.1 | 17 - 18 | 2409 | 84 | |
GenBank mRNA | AK067082.1 | - | 17 - 18 | 2272 | 84 | |
KT199367.1 | - | 17 - 18 | 2173 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
