Assay Details
Gene Symbol: LOC4344192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791726.1 | XP_015647212.1 | 19 - 20 | 2681 | 73 | |
GenBank mRNA | KT199367.1 | - | 19 - 20 | 2445 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
