Assay Details
| Gene Symbol: LOC4344192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791726.1 | XP_015647212.1 | 1 - 2 | 388 | 77 | |
| GenBank mRNA | AK067082.1 | - | 1 - 2 | 251 | 77 | |
| KT199367.1 | - | 1 - 2 | 152 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015791726.1 transcripts.
Set Membership: |
|

LOC4344192
Genomic Map