Assay Details
| Gene Symbol: LOC4344192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791726.1 | XP_015647212.1 | 22 - 23 | 3066 | 62 | |
| GenBank mRNA | AK067082.1 | - | 22 - 23 | 2928 | 62 | |
| KT199367.1 | - | 22 - 23 | 2830 | 62 | ||
More Information
Set Membership: |
|

LOC4344192
Genomic Map