Assay Details
| Gene Symbol: LOC4344192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791726.1 | XP_015647212.1 | 9 - 10 | 1466 | 66 | |
| GenBank mRNA | AK067082.1 | - | 9 - 10 | 1329 | 66 | |
| KT199367.1 | - | 9 - 10 | 1230 | 66 | ||
More Information
Set Membership: |
|

LOC4344192
Genomic Map