Assay Details
| Gene Symbol: LOC4344227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789626.2 | XP_015645112.1 | 5 - 6 | 720 | 121 | |
| XM_015789627.2 | XP_015645113.1 | 5 - 6 | 363 | 121 | ||
| GenBank mRNA | AK072772.1 | - | 4 - 5 | 1225 | 121 | |
| AK108593.1 | - | 5 - 6 | 520 | 121 | ||
More Information
Set Membership: |
|

LOC4344227
Genomic Map