Assay Details
| Gene Symbol: LOC4344227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789626.2 | XP_015645112.1 | 6 - 7 | 785 | 133 | |
| XM_015789627.2 | XP_015645113.1 | 6 - 7 | 428 | 133 | ||
| GenBank mRNA | AK108593.1 | - | 6 - 7 | 585 | 133 | |
More Information
Set Membership: |
|

LOC4344227
Genomic Map