Assay Details
| Gene Symbol: LOC4344685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794626.2 | XP_015650112.1 | 2 - 3 | 1501 | 87 | |
| GenBank mRNA | AK070325.1 | - | 2 - 3 | 1502 | 87 | |
| AK099134.1 | - | 2 - 3 | 1482 | 87 | ||
More Information
Set Membership: |
|

LOC4344685
Genomic Map