Assay Details
| Gene Symbol: LOC4344685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794626.2 | XP_015650112.1 | 8 - 9 | 2208 | 145 | |
| GenBank mRNA | AK070325.1 | - | 8 - 9 | 2209 | 145 | |
| AK099134.1 | - | 8 - 9 | 2188 | 145 | ||
More Information
Set Membership: |
|

LOC4344685
Genomic Map