Assay Details
| Gene Symbol: LOC4344801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794249.2 | XP_015649735.1 | 11 - 12 | 3309 | 69 | |
| GenBank mRNA | AK101719.1 | - | 11 - 12 | 3314 | 69 | |
More Information
Set Membership: |
|

LOC4344801
Genomic Map