Assay Details
| Gene Symbol: LOC4344937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794913.2 | XP_015650399.1 | 2 - 3 | 917 | 77 | |
| GenBank mRNA | AB126089.1 | - | 2 - 3 | 889 | 77 | |
| AK105258.1 | - | 2 - 3 | 923 | 77 | ||
More Information
Set Membership: |
|

LOC4344937
Genomic Map