Assay Details
Gene Symbol: LOC4346386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015757081.2 | XP_015612567.1 | 1 - 2 | 189 | 92 | |
GenBank mRNA | AK067482.1 | - | 1 - 2 | 158 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
