Assay Details
Gene Symbol: LOC4346386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015757081.2 | XP_015612567.1 | 7 - 8 | 588 | 65 | |
GenBank mRNA | AK067482.1 | - | 7 - 8 | 557 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
