Assay Details
Gene Symbol: LOC4347368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756205.2 | XP_015611691.1 | 13 - 14 | 1105 | 108 | |
GenBank mRNA | AK068464.1 | - | 13 - 14 | 1101 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
