Assay Details
Gene Symbol: LOC4347368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756205.2 | XP_015611691.1 | 16 - 17 | 1394 | 109 | |
GenBank mRNA | AK068464.1 | - | 16 - 17 | 1390 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
