Assay Details
Gene Symbol: LOC4347368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756205.2 | XP_015611691.1 | 18 - 19 | 1550 | 108 | |
GenBank mRNA | AK068464.1 | - | 18 - 19 | 1546 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
