Assay Details
Gene Symbol: LOC4347517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015755338.1 | XP_015610824.1 | 5 - 6 | 593 | 90 | |
GenBank mRNA | AK072867.1 | - | 5 - 6 | 589 | 90 | |
U78892.1 | - | 5 - 6 | 587 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
