Assay Details
| Gene Symbol: LOC4327093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766105.2 | XP_015621591.1 | 1 | 1950 | 86 | |
| GenBank mRNA | AK065589.1 | - | 1 | 1849 | 86 | |
| AK069681.1 | - | 1 | 993 | 86 | ||
| AK071552.1 | - | 1 | 1729 | 86 | ||
More Information
Set Membership: |
|

LOC4327093
Genomic Map