Assay Details
Gene Symbol: LOC4327789 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015778945.2 | XP_015634431.1 | 1 - 2 | 339 | 85 | |
GenBank mRNA | AK062426.1 | - | 1 - 2 | 335 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
