Assay Details
| Gene Symbol: LOC4325129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765904.2 | XP_015621390.1 | 1 - 2 | 335 | 82 | |
| GenBank mRNA | AK061583.1 | - | 1 - 2 | 335 | 82 | |
| AK102117.1 | - | 1 - 2 | 336 | 82 | ||
| AK243051.1 | - | 1 - 2 | 329 | 82 | ||
More Information
Set Membership: |
|

LOC4325129
Genomic Map