Assay Details
Gene Symbol: LOC4325451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766402.2 | XP_015621888.1 | 1 - 2 | 1263 | 66 | |
GenBank mRNA | AK059266.1 | - | 1 - 2 | 1045 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
