Assay Details
Gene Symbol: LOC4325962 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776028.2 | XP_015631514.1 | 4 - 5 | 523 | 151 | |
XM_015776037.2 | XP_015631523.1 | 4 - 5 | 523 | 151 | ||
XM_015776045.2 | XP_015631531.1 | 4 - 5 | 485 | 151 | ||
GenBank mRNA | AK111570.1 | - | 4 - 5 | 487 | 151 | |
DQ201199.1 | - | 3 - 4 | 229 | 151 | ||
FJ693712.1 | - | 3 - 4 | 295 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() |
