Assay Details
Gene Symbol: LOC4326574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766584.1 | XP_015622070.1 | 16 - 17 | 2007 | 142 | |
GenBank mRNA | AK099822.1 | - | 16 - 17 | 2001 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() |
