Assay Details
Gene Symbol: LOC4326574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766584.1 | XP_015622070.1 | 2 - 3 | 221 | 79 | |
XM_015766588.2 | XP_015622074.1 | 1 - 2 | 68 | 79 | ||
GenBank mRNA | AK099822.1 | - | 2 - 3 | 215 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015766584.1, XM_015766588.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
