Assay Details
Gene Symbol: LOC4324432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772170.2 | XP_015627656.1 | 1 - 2 | 549 | 119 | |
XM_015772177.2 | XP_015627663.1 | 1 - 2 | 549 | 119 | ||
GenBank mRNA | AK121474.1 | - | 1 - 2 | 466 | 119 | |
AK288900.1 | - | 1 - 2 | 319 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
