Assay Details
Gene Symbol: LOC4324993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756658.2 | XP_015612144.1 | 2 - 3 | 525 | 108 | |
XM_015756665.2 | XP_015612151.1 | 2 - 3 | 1246 | 108 | ||
GenBank mRNA | AK102849.1 | - | 2 - 3 | 1149 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
