Assay Details
Gene Symbol: LOC4324993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756658.2 | XP_015612144.1 | 3 - 4 | 873 | 76 | |
XM_015756665.2 | XP_015612151.1 | 3 - 4 | 1594 | 76 | ||
GenBank mRNA | AK102849.1 | - | 3 - 4 | 1497 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
