Assay Details
| Gene Symbol: LOC4324962 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766475.2 | XP_015621961.1 | 1 - 2 | 615 | 109 | |
| GenBank mRNA | AK060500.1 | - | 1 - 2 | 616 | 109 | |
| AK120887.1 | - | 2 - 3 | 260 | 109 | ||
More Information
Set Membership: |
|

LOC4324962
Genomic Map