Assay Details
| Gene Symbol: LOC4325027 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758323.2 | XP_015613809.1 | 1 - 2 | 175 | 99 | |
| GenBank mRNA | AK062270.1 | - | 1 - 2 | 96 | 99 | |
| AK073770.1 | - | 1 - 2 | 142 | 99 | ||
| AK104719.1 | - | 1 - 2 | 96 | 99 | ||
| EF122473.1 | - | 1 - 2 | 131 | 99 | ||
| GQ848028.1 | - | 1 - 2 | 79 | 99 | ||
| JN208224.1 | - | 1 - 2 | 73 | 99 | ||
| KT862205.1 | - | 1 - 2 | 73 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015758323.2 transcripts.
Set Membership: |
|

LOC4325027
Genomic Map