Assay Details
Gene Symbol: LOC9271802 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015793086.2 | XP_015648572.1 | 2 - 3 | 1205 | 65 | |
GenBank mRNA | AK064782.1 | - | 2 - 3 | 635 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
