Assay Details
Gene Symbol: LOC4326830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787050.2 | XP_015642536.1 | 1 | 1262 | 79 | |
GenBank mRNA | AK100995.1 | - | 1 | 1247 | 79 | |
AK120801.1 | - | 1 | 1243 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
