Assay Details
| Gene Symbol: LOC107278964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757024.1 | XP_015612510.1 | 5 - 6 | 1648 | 90 | |
| XM_015757033.1 | XP_015612519.1 | 5 - 6 | 455 | 90 | ||
| GenBank mRNA | AK241684.1 | - | 5 - 6 | 1599 | 90 | |
More Information
Set Membership: |
|

LOC107278964
Genomic Map