Assay Details
Gene Symbol: LOC4327993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771048.2 | XP_015626534.1 | 1 - 2 | 156 | 93 | |
GenBank mRNA | AK070603.1 | - | 1 - 2 | 109 | 93 | |
AK101537.1 | - | 1 - 2 | 88 | 93 | ||
GU120333.1 | - | 1 - 2 | 109 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015771048.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
