Assay Details
Gene Symbol: LOC4327993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771048.2 | XP_015626534.1 | 2 - 3 | 363 | 115 | |
GenBank mRNA | AK070603.1 | - | 2 - 3 | 316 | 115 | |
AK101537.1 | - | 2 - 3 | 295 | 115 | ||
GU120333.1 | - | 2 - 3 | 316 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
