Assay Details
Gene Symbol: LOC4327993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771048.2 | XP_015626534.1 | 5 - 6 | 721 | 78 | |
GenBank mRNA | AK101537.1 | - | 5 - 6 | 653 | 78 | |
GU120333.1 | - | 5 - 6 | 674 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
