Assay Details
Gene Symbol: LOC4327993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771048.2 | XP_015626534.1 | 9 - 10 | 1454 | 85 | |
GenBank mRNA | AK101537.1 | - | 9 - 10 | 1386 | 85 | |
GU120333.1 | - | 9 - 10 | 1407 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
