Assay Details
Gene Symbol: LOC4328210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770954.2 | XP_015626440.1 | 16 - 17 | 1825 | 116 | |
XM_015770955.2 | XP_015626441.1 | 16 - 17 | 1827 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
