Assay Details
Gene Symbol: LOC4328210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770954.2 | XP_015626440.1 | 20 - 21 | 2408 | 80 | |
XM_015770955.2 | XP_015626441.1 | 20 - 21 | 2410 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
